missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol), Reagents
Specifications
Specifications
| Melting Point | -80°C |
| Color | Colorless |
| Quantity | 500 mL |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Specific Gravity | 0.785 |
| Boiling Point | 72°C |
| Percent Purity | Min. 99.5% |
| Grade | ACS |
| Chemical Name or Material | Reagent Alcohol, ACS Reagent, Absolute (Anhydrous Denatured Ethanol) |