missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Ready-To-Go™ GenomiPhi HY DNA Amplification Kit
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60++g yield from nanogram amounts of DNA sample.
$893.45 - $2422.00
Specifications
| Content And Storage | Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons |
|---|---|
| Description | illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample. |
| For Use With (Application) | Genomic DNA Amplification |
| Size | > 10 kb |
| Test Time | 4 h |
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity | |||||
|
45-003-104
|
Cytiva
25660324 |
24 |
Each for $893.45
|
|
|||||
|
45-003-105
|
Cytiva
25660396 |
96 |
Each for $2,422.00
|
|
|||||
Description
illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit is part of the Phi29 DNA polymerase family of products from Cytiva.
- Representative amplification of the whole genome.
- Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
- Quick and simple automation-friendly protocol; no thermal cycler required.
- Shipping and storage at room temperature provides easy access and reduced carbon footprint.
- Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
Specifications
| Store the kit at ambient temperature.Once opened, completely reseal the pouch containingthe Ready-To-Go GenomiPhi V3 DNA amplification kit cakes. To ensure maximum shelf life and optimal performance, storage in a desiccator is recommended.Store recons | |
| Genomic DNA Amplification | |
| 4 h |
| illustra Ready-To-Go GenomiPhi HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample. | |
| > 10 kb |