missing translation for 'onlineSavingsMsg'
Learn More

RD Plastics Reclosable Specimen Transport Bag

Light duty and available in both two or three wall construction

$54.53

Specifications

Color Clear
For Use With (Application) For specimen transport
Thickness (English) 2 mil
Product Type Reclosable Specimen Transport Bag
Thickness (Metric) 0.279 mm
View More Specs

Products 4
Catalog Number Mfr. No. Dimensions (L x W) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Includes Price Quantity & Availability  
23700207
View Documents Promotion Details
RD Plastics
Q72
6 x 9 in. 2-wall Construction, Storage Instructions
Case of 1000 Each for $54.53
Only null left
 
23700236
View Documents Promotion Details
RD Plastics
Q512
6 x 4 in. 2-wall Construction, Storage Instructions
Case of 1000 Each for $54.53
Only null left
 
23700208
View Documents Promotion Details
RD Plastics
RDB101
10 x 8 in. Absorbent Pad
Case of 500 Each for $54.53
Only null left
 
01800102
View Documents Promotion Details
RD Plastics
Q190
10 x 10 in. 2-wall Construction, Storage Instructions
Case of 1000 Each for $54.53
Only null left
 
Description

Description

 

  • Specimen transport
  • Two wall or three wall
  • Some have flap cover to protect papers
  • Biohazard print
  • Score line for easy opening
  • Absobent pad

 

Specifications

Specifications

Clear
2 mil
0.279 mm
For specimen transport
Reclosable Specimen Transport Bag
Videos
SDS
Documents

Documents