missing translation for 'onlineSavingsMsg'
Learn More

RD Plastics Reclosable Specimen Transport Bag

Catalog No. 23700207
Change view
Click to view available options
Dimensions (L x W):
6 x 4 in.
6 x 9 in.
10 x 8 in.
10 x 10 in.
Includes:
2-wall Construction, Storage Instructions
Absorbent Pad
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W) Includes
23700207 6 x 9 in. 2-wall Construction, Storage Instructions
23700236 6 x 4 in. 2-wall Construction, Storage Instructions
23700208 10 x 8 in. Absorbent Pad
01800102 10 x 10 in. 2-wall Construction, Storage Instructions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23700207 Supplier RD Plastics Supplier No. Q72

Light duty and available in both two or three wall construction

 

  • Specimen transport
  • Two wall or three wall
  • Biohazard print
  • Score line for easy opening
  • Absobent pad

 

Specifications

Closure Type Reclosable
Color Clear
For Use With (Application) For specimen transport
Includes 2-wall Construction, Storage Instructions
Thickness (English) 2 mil
Dimensions (L x W) 6 x 9 in.
Length (English) 8 in.
Width (English) 10 in.
Product Type Reclosable Specimen Transport Bag
Length (Metric) 20.3 cm
Width (Metric) 25.4 cm
Thickness (Metric) 0.279 mm
Show More Show Less