missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RD Plastics Reclosable Specimen Transport Bag
Light duty and available in both two or three wall construction
Supplier: RD Plastics Q512
Description
- Specimen transport
- Two wall or three wall
- Biohazard print
- Score line for easy opening
Chemical Identifiers
| Clear | |
| 2 mil | |
| 0.279 mm |
| For specimen transport | |
| Reclosable Specimen Transport Bag |
Specifications
| Reclosable Zip | |
| For specimen transport | |
| 2 mil | |
| 6 in. | |
| Reclosable Specimen Transport Bag | |
| 10.1 cm |
| Clear | |
| 2-wall Construction, Storage Instructions | |
| 6 x 4 in. | |
| 4 in. | |
| 15.2 cm | |
| 0.279 mm |