missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCOR1 Monoclonal Antibody (K40046_9A7), Invitrogen™
Mouse Monoclonal Antibody
$1,740.58 - $2,926.56
Specifications
| Antigen | RCOR1 |
|---|---|
| Clone | K40046_9A7 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: RKQKREREES EDELEEANGN NPIDIEVDQN KESKKEVPPT ETVPQVKKEK HSTQAKNRAK RKPPKGMFLS QEDVEAVSAN ATAATTVLRQ LDMELVSVKR QIQNIKQTNS ALKEKLDGGI EPYRLPEVIQ KCNARWTTEE QLLAVQAIRK YGRDFQAISD VIGNKSVVQV KNFFVNYRRR FNIDEVLQEW EAEHGKEETN GPSNQKPVKS PDNSIKMPEE
RCOR1 Monoclonal antibody specifically detects RCOR1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| RCOR1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 5730409O11; 6720480E22Rik; AU042633; COREST; D12Wsu95e; im:6904264; KIAA0071; LOC102554884; mKIAA0071; Protein CoREST; RCOR; Rcor1; REST corepressor 1; REST corepressor 1-like; Rocr1; si:dkey-46n18.7; zgc:158607 | |
| Recombinant human RCOR1 (immunogen range is 251-470). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| RCOR1 |
| K40046_9A7 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UKL0 | |
| 23186 | |
| IgG1 | |
| Protein A | |
| Antibody |