missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RCOR1 Monoclonal Antibody (K40046_9A7), Invitrogen™

Catalog No. MA556351
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556351 200 μL
MA555971 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556351 Supplier Invitrogen™ Supplier No. MA556351
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: RKQKREREES EDELEEANGN NPIDIEVDQN KESKKEVPPT ETVPQVKKEK HSTQAKNRAK RKPPKGMFLS QEDVEAVSAN ATAATTVLRQ LDMELVSVKR QIQNIKQTNS ALKEKLDGGI EPYRLPEVIQ KCNARWTTEE QLLAVQAIRK YGRDFQAISD VIGNKSVVQV KNFFVNYRRR FNIDEVLQEW EAEHGKEETN GPSNQKPVKS PDNSIKMPEE

RCOR1 is a functional corepressor required for regulation of neural-specific gene expression. The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression. The RCOR gene encodes a functional corepressor required for regulation of neural-specific gene expression.

Specifications

Antigen RCOR1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K40046_9A7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene RCOR1
Gene Accession No. Q9UKL0
Gene Alias 5730409O11; 6720480E22Rik; AU042633; COREST; D12Wsu95e; im:6904264; KIAA0071; LOC102554884; mKIAA0071; Protein CoREST; RCOR; Rcor1; REST corepressor 1; REST corepressor 1-like; Rocr1; si:dkey-46n18.7; zgc:158607
Gene Symbols RCOR1
Host Species Mouse
Immunogen Recombinant human RCOR1 (immunogen range is 251-470).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23186
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less