missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat IL-13 (113 aa) Recombinant Protein, PeproTech®
Recombinant Protein
$132.00 - $8070.00
Specifications
| Accession Number | P42203 |
|---|---|
| For Use With (Application) | ELISA, Functional Assay, Western Blot |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 116553 |
| Molecular Weight (g/mol) | 12.3 kDa |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
40016L100UG
|
Gibco™
40016L100UG |
100 μg |
Each for $1,614.59
|
|
|||||
40016L10UG
|
Gibco™
40016L10UG |
10 μg |
Each for $302.74
|
|
|||||
40016L1MG
|
Gibco™
40016L1MG |
1 mg |
Each for $8,070.00
|
|
|||||
40016L250UG
|
Gibco™
40016L250UG |
250 μg |
Each for $2,825.00
|
|
|||||
40016L2UG
|
Gibco™
LS40016L2UG |
2 μg |
Each for $132.00
|
|
|||||
40016L500UG
|
Gibco™
40016L500UG |
500 μg |
Each for $4,835.00
|
|
|||||
Description
Recombinant Rat IL-13 a 12.3 kDa protein consisting of 113 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
IL-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells, and is involved in regulating inflammatory and immune responses. IL-13 down-regulates macrophage activity, thereby inhibiting the production of pro-inflammatory cytokines and chemokines, and is involved in allergen-induced asthma through mechanisms independent of IgE and eosinophils. IL-13 also regulates several stages of B cell maturation and differentiation, up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. Structurally, IL-13 closely resembles IL-3, IL-4, IL-5 and GM-CSF. The gene encoding IL-13, IL3, IL5, IL4, and CSF2 form a cytokine gene cluster on chromosome 5q, with the IL-13 gene particularly close to IL4.Specifications
| P42203 | |
| protein with no preservative | |
| 12.3 kDa | |
| E. coli | |
| <1 EU/ μg | |
| IL-13 | |
| The ED50 was determined by the dose-dependent proliferation of TF-1 cells is ≤ 5.0 ng/ml, corresponding to a specific activity of ≥ 2 x 10^5 units/mg | |
| Recombinant | |
| -20°C | |
| Lyophilized |
| ELISA, Functional Assay, Western Blot | |
| 116553 | |
| Rat IL-13 (113 aa) | |
| RUO | |
| ALRH; ALRH antibody; BHR1; il 13; Il13; Il-13; IL-13; cytokine; ILN; Interleukin; interleukin 13; Interleukin13; interleukin-13; interleukin-13 precursor; L13; MGC116786; MGC116788; MGC116789; NC30; NC30 antibody; P600; P600 antibody; T-cell activation protein P600 | |
| Il13 | |
| Unconjugated | |
| TPGPVRRSTS PPVALRELIE ELSNITQDQK TSLCNSSMVW SVDLTAGGFC AALESLTNIS SCNAIHRTQR ILNGLCNQKA SDVASSPPDT KIEVAQFISK LLNYSKQLFR YGH | |
| E. coli | |
| ≥ 98% by SDS-PAGE gel and HPLC analyses. |