Learn More
Thermo Scientific™ MarqMetrix Replacement Bioreactor BallProbe
Description
- Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases and most organic solvents including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane and acetonitrile
Specifications
Specifications
| Product Type | Replacement Bioreactor Ball Probe |
| For Use With (Equipment) | MarqMetrix All-In-One Process Raman Analyzer |