missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: Lithium Heparin
No scooping needed, allowing for high-quality, tissue-free samples
$221.07 - $2,056.50
Specifications
| Product Type | Chemistry/Plasma/Whole Blood Tube |
|---|---|
| Material | Plastic |
| Sterility | Non-sterile |
| Closure Type | Standard Pierceable Cap |
| Closure Included | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Additive | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Additive | Price | Quantity & Availability | ||||
|
1491564
|
RAM Scientific
07 6101 |
125 μL | Lithium Heparin Mini |
Pack of 50 Each for $231.32
Case of 10 Pack for $2,313.20
|
|
||||
|
1491566
|
RAM Scientific
07 7251 |
300 μL | Lithium Heparin |
Pack of 50 Each for $221.07
Case of 10 Pack for $2,210.70
|
|
||||
|
1491565
|
RAM Scientific
07 7250 |
200 μL | Lithium Heparin |
Pack of 50 Each for $221.07
Case of 10 Pack for $2,210.70
|
|
||||
|
1491569
|
RAM Scientific
07 7221 |
300 μL | Lithium Heparin with Gel |
Case of 500 Each for $2,056.50
|
|
||||
|
1491568
|
RAM Scientific
07 7220 |
200 μL | Lithium Heparin with Gel |
Pack of 50 Each for $232.39
Case of 10 Pack for $2,323.90
|
|
||||
Description
No scooping needed, allowing for high-quality, tissue-free samples
- Pierceable attached cap with self-sealing versions
- Anticoagulant coated capillary and tube to prevent clotting
- Expiration date and lot number on tube for quality control
Specifications
| Chemistry/Plasma/Whole Blood Tube | |
| Non-sterile | |
| Yes |
| Plastic | |
| Standard Pierceable Cap | |
| Green |