missing translation for 'onlineSavingsMsg'
Learn More

RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: Lithium Heparin

Catalog No. 1491569
Change view
Click to view available options
Capacity (Metric):
200 μL
300 μL
Additive:
Lithium Heparin with Gel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Additive
1491569 300 μL Lithium Heparin with Gel
1491568 200 μL Lithium Heparin with Gel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 1491569 Supplier RAM Scientific Supplier No. 07 7221

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Pre-assembled capillary blood collection device with a capillary tube, attached pierceable cap, and a microtube

No scooping needed, allowing for high-quality, tissue-free samples

No scooping needed, allowing for high-quality, tissue-free samples

  • 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
  • Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
  • Capillary straw and microtube are coated with anticoagulant Lithium Heparin (Green cap) to prevent clotting
  • Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
  • Lot Number, Expiration Date, and fill lines are printed directly on the microtube
  • A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
  • For plasma determinations in chemistry
  • Small sample sizes, great for pediatrics and neonates
  • Optional gel separator serves as a barrier between blood and serum after centrifugation
  • Available in a variety of sizes and draw volumes, with or without gel separator
  • Optional amber microtube ensures light-sensitive sample integrity providing protection for testing (ex. Bilirubin)

Recommended Storage

Store at room temperature. Keep dry. Keep away from sunlight.

Specifications

Color Clear
Product Type Chemistry/Plasma/Whole Blood Tube
Additive Lithium Heparin with Gel
Material Plastic
Sterility Non-sterile
Closure Type Standard Pierceable Cap
Capacity (Metric) 300 μL
Closure Included Yes
Closure Color Green
For Use With (Application) Intended for chemistry determinations that require plasma or whole blood samples. Gel separator serves as a barrier between blood and serum after centrifugation.