missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAM Scientific Safe-T-Fill™ Capillary Blood Collection Systems: Lithium Heparin
Description
Includes
Pre-assembled capillary blood collection device with a capillary tube, attached pierceable cap, and a microtube
No scooping needed, allowing for high-quality, tissue-free samples
- 100% plastic tubes collect microliter volume capillary blood samples from finger and heel sticks
- Blood samples are drawn into the microtube through capillary action, reducing hemolysis, tissue contamination, and re-draws. No "scooping" or "milking" required
- Capillary straw and microtube are coated with anticoagulant Lithium Heparin (Green cap) to prevent clotting
- Attached cap is color-coded to designate the type of additive that is in both the capillary tube and the microtube
- Lot Number, Expiration Date, and fill lines are printed directly on the microtube
- A complete end-to-end fill of the capillary tube accurately measures the stated fill volume
- For plasma determinations in chemistry
- Small sample sizes, great for pediatrics and neonates
- Optional gel separator serves as a barrier between blood and serum after centrifugation
- Available in a variety of sizes and draw volumes, with or without gel separator
- Optional amber microtube ensures light-sensitive sample integrity providing protection for testing (ex. Bilirubin)
Recommended Storage
Store at room temperature. Keep dry. Keep away from sunlight.
Specifications
Specifications
| Color | Clear |
| Product Type | Chemistry/Plasma/Whole Blood Tube |
| Additive | Lithium Heparin |
| Material | Plastic |
| Sterility | Non-sterile |
| Closure Type | Standard Pierceable Cap |
| Capacity (Metric) | 300 μL |
| Closure Included | Yes |
| Closure Color | Green |
| For Use With (Application) | Intended for chemistry determinations that require plasma or whole blood samples. |