missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole Sulfide 98.0+%, TCI America™

Catalog No. p-7135694
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0098200MG 200 mg
R00981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0098200MG Supplier TCI America Supplier No. R0098200MG

Chemical Identifiers

CAS 117977-21-6
Molecular Formula C18H21N3O2S
Molecular Weight (g/mol) 343.45
MDL Number MFCD08063845
InChI Key BSXAHDOWMOSVAP-UHFFFAOYSA-N
Synonym 2-[[4-(3-Methoxypropoxy)-3-methyl-2-pyridyl]methylthio]benzimidazole
PubChem CID 9949996
IUPAC Name 2-({[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methyl}sulfanyl)-1H-1,3-benzodiazole
SMILES COCCCOC1=C(C)C(CSC2=NC3=CC=CC=C3N2)=NC=C1

Specifications

Melting Point 116°C
Color White-Yellow
Quantity 200 mg
Formula Weight 343.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Rabeprazole Sulfide