missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-TolBINAP 98.0+%, TCI America™

Catalog No. p-7137607
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31521G 1 g
T31525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31521G Supplier TCI America Supplier No. T31521G
Only null left

Chemical Identifiers

CAS 99646-28-3
Molecular Formula C48H40P2
Molecular Weight (g/mol) 678.796
MDL Number MFCD01311709
InChI Key IOPQYDKQISFMJI-UHFFFAOYSA-N
Synonym r-tol-binap, s-tol-binap, s---2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, 2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-t-binap, rac-tol-binap, r-+-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, tol-binap, s-2,2'-bis di-p-tolylphosphino-1,1'-binaphthyl, s-p-tol-binap
PubChem CID 4189906
IUPAC Name [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C

Specifications

Melting Point 258°C
Color White-Yellow
Quantity 1 g
Formula Weight 678.80
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-TolBINAP