accessibility menu, dialog, popup

UserName

H3CPCH3PH3CCH3(?)

CAS RN 99646-28-3

IUPAC Name: {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane
Synonyms: {2'-[bis(4-methylphenyl)phosphanyl]-[1,1'-binaphthalen]-2-yl}bis(4-methylphenyl)phosphane
Molecular Weight (g/mol): 678.8
Molecular Formula: C48H40P2
InChi Key: IOPQYDKQISFMJI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)