missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-Propylene Oxide 98.0+%, TCI America™
Supplier: TCI America P139625ML
Specifications
| (R)-(+)-Propylene Oxide | |
| Colorless | |
| C3H6O | |
| 25 mL | |
| r-+-propylene oxide, r-propylene oxide, 2r-2-methyloxirane, r-2-methyloxirane, r-methyloxirane, +-propylene oxide, r-epoxypropane, +-methyloxirane, r-+-1,2-epoxypropane, r-1,2-epoxypropane | |
| CC1CO1 | |
| 58.08 | |
| CHEBI:28985 | |
| ≥98.0% (GC) |
| 15448-47-2 | |
| 34°C | |
| MFCD00066211 | |
| 1280 | |
| GOOHAUXETOMSMM-GSVOUGTGSA-N | |
| (2R)-2-methyloxirane | |
| 146261 | |
| 58.08 | |
| Liquid |
Chemical Identifiers
| 15448-47-2 | |
| 58.08 | |
| GOOHAUXETOMSMM-GSVOUGTGSA-N | |
| 146261 | |
| (2R)-2-methyloxirane |
| C3H6O | |
| MFCD00066211 | |
| r-+-propylene oxide, r-propylene oxide, 2r-2-methyloxirane, r-2-methyloxirane, r-methyloxirane, +-propylene oxide, r-epoxypropane, +-methyloxirane, r-+-1,2-epoxypropane, r-1,2-epoxypropane | |
| CHEBI:28985 | |
| CC1CO1 |
Safety and Handling
EINECSNumber : (2)-0219
RTECSNumber : UJ2650000
TSCA : No
Recommended Storage : Refrigerator