missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Propylene Oxide 98.0+%, TCI America™

Catalog No. p-7135115
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
P13965ML 5 mL
P139625ML 25 mL
2 options

Catalog No. P13965ML

Supplier: TCI America P13965ML

Only null left

Chemical Identifiers

CAS 15448-47-2
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00066211
InChI Key GOOHAUXETOMSMM-GSVOUGTGSA-N
Synonym r-+-propylene oxide, r-propylene oxide, 2r-2-methyloxirane, r-2-methyloxirane, r-methyloxirane, +-propylene oxide, r-epoxypropane, +-methyloxirane, r-+-1,2-epoxypropane, r-1,2-epoxypropane
PubChem CID 146261
ChEBI CHEBI:28985
IUPAC Name (2R)-2-methyloxirane
SMILES CC1CO1

Specifications

Color Colorless
Boiling Point 34°C
Quantity 5 mL
UN Number 1280
Formula Weight 58.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-Propylene Oxide
EINECSNumber (2)-0219
RTECSNumber UJ2650000
TSCA No
Recommended Storage Refrigerator