accessibility menu, dialog, popup

UserName

CAS RN 15448-47-2

IUPAC Name: (2R)-2-methyloxirane
Synonyms: (+)-propylene oxide (R)-epoxypropane (+)-methyloxirane
Molecular Weight (g/mol): 58.08
Molecular Formula: C3H6O
InChi Key: GOOHAUXETOMSMM-GSVOUGTGSA-N
SMILES: C[C@@H]1CO1
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)