missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1865903

 View more versions of this product

Catalog No. AAL1865903

Only null left
Explore available promotions

Description

Description

(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine is a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Store under inert gas. Incompatible with oxidizing agents.
Specifications

Specifications

(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine
36°C to 40°C
MFCD06795639
UN3259
Air sensitive
Insoluble in water.
−90° (c=1 in Methanol)
2,2-dimethyl-N-[(1R)-1-phenyl-2-piperidin-1-ylethyl]propan-1-amine
7577799
97%
153837-28-6
C18H30N2
1 g
6957993
r-2,2-dimethyl-n-1-phenyl-2-piperidin-1-yl ethyl propan-1-amine, r---n-neopentyl-1-phenyl-2-1-piperidinyl ethylamine, 1r-1-phenyl-2-piperidylethyl 2,2-dimethylpropyl amine, 2,2-dimethylpropyl 1r-1-phenyl-2-piperidin-1-yl ethyl amine, 1r-n-2,2-dimethylpropyl-1-phenyl-2-piperidinoethanamine, 2,2-dimethyl-n-1r-1-phenyl-2-piperidin-1-ylethyl propan-1-amine
RUWFXOINQANLGF-KRWDZBQOSA-N
CC(C)(C)CNC(CN1CCCCC1)C2=CC=CC=C2
274.452
274.45

Chemical Identifiers

153837-28-6
274.452
RUWFXOINQANLGF-KRWDZBQOSA-N
7577799
CC(C)(C)CNC(CN1CCCCC1)C2=CC=CC=C2
C18H30N2
MFCD06795639
r-2,2-dimethyl-n-1-phenyl-2-piperidin-1-yl ethyl propan-1-amine, r---n-neopentyl-1-phenyl-2-1-piperidinyl ethylamine, 1r-1-phenyl-2-piperidylethyl 2,2-dimethylpropyl amine, 2,2-dimethylpropyl 1r-1-phenyl-2-piperidin-1-yl ethyl amine, 1r-n-2,2-dimethylpropyl-1-phenyl-2-piperidinoethanamine, 2,2-dimethyl-n-1r-1-phenyl-2-piperidin-1-ylethyl propan-1-amine
2,2-dimethyl-N-[(1R)-1-phenyl-2-piperidin-1-ylethyl]propan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only