accessibility menu, dialog, popup

UserName

CAS RN 153837-28-6

IUPAC Name: (2,2-dimethylpropyl)[1-phenyl-2-(piperidin-1-yl)ethyl]amine
Synonyms: (2,2-dimethylpropyl)[1-phenyl-2-(piperidin-1-yl)ethyl]amine
Molecular Weight (g/mol): 274.45
Molecular Formula: C18H30N2
InChi Key: RUWFXOINQANLGF-UHFFFAOYNA-N
SMILES: CC(C)(C)CNC(CN1CCCCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results