missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine, 97%, Thermo Scientific Chemicals

Catalog No. p-7047311
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1865903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1865903 Supplier Thermo Scientific Chemicals Supplier No. L1865903

(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine is a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Store under inert gas. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 153837-28-6
Molecular Formula C18H30N2
Molecular Weight (g/mol) 274.452
MDL Number MFCD06795639
InChI Key RUWFXOINQANLGF-KRWDZBQOSA-N
Synonym r-2,2-dimethyl-n-1-phenyl-2-piperidin-1-yl ethyl propan-1-amine, r---n-neopentyl-1-phenyl-2-1-piperidinyl ethylamine, 1r-1-phenyl-2-piperidylethyl 2,2-dimethylpropyl amine, 2,2-dimethylpropyl 1r-1-phenyl-2-piperidin-1-yl ethyl amine, 1r-n-2,2-dimethylpropyl-1-phenyl-2-piperidinoethanamine, 2,2-dimethyl-n-1r-1-phenyl-2-piperidin-1-ylethyl propan-1-amine
PubChem CID 7577799
IUPAC Name 2,2-dimethyl-N-[(1R)-1-phenyl-2-piperidin-1-ylethyl]propan-1-amine
SMILES CC(C)(C)CNC(CN1CCCCC1)C2=CC=CC=C2

Specifications

Melting Point 36°C to 40°C
Quantity 1 g
UN Number UN3259
Beilstein 6957993
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Optical Rotation −90° (c=1 in Methanol)
Formula Weight 274.45
Percent Purity 97%
Chemical Name or Material (R)-(-)-N-Neopentyl-1-phenyl-2-(1-piperidinyl)ethylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only