missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-Allylglycine, 95%, 98% ee, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 362840010

 View more versions of this product

Catalog No. AC362840010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

(R)-N-BOC-Allylglycine
170899-08-8
100.0
Authentic
Glass bottle
MFCD01320882
-9.5° to -12.5° (20°C, 589nm) (c=1, methanol)
BUPDPLXLAKNJMI-SSDOTTSWSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
638723
95%,98% ee
95%, (98% e.e.)
94.0
97.5% min.
94% min. (GC)
C10H17NO4
1g
boc-d-allylglycine, r-2-tert-butoxycarbonyl amino pent-4-enoic acid, boc-alpha-allyl-d-gly, boc-d-allyglycine.dcha, 2r-2-tert-butoxycarbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, boc-d-allylglycine.dcha, d-boc-allylglycine, boc-d-allyglycine?dcha, boc-d-allyglycine?cha
CC(C)(C)OC(=O)NC(CC=C)C(=O)O
215.25
215.25

Chemical Identifiers

170899-08-8
215.25
BUPDPLXLAKNJMI-SSDOTTSWSA-N
638723
CC(C)(C)OC(=O)NC(CC=C)C(=O)O
C10H17NO4
MFCD01320882
boc-d-allylglycine, r-2-tert-butoxycarbonyl amino pent-4-enoic acid, boc-alpha-allyl-d-gly, boc-d-allyglycine.dcha, 2r-2-tert-butoxycarbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, boc-d-allylglycine.dcha, d-boc-allylglycine, boc-d-allyglycine?dcha, boc-d-allyglycine?cha
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

GHS Signal Word: Warning

TSCA : TSCA

Recommended Storage : 4°C

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only