accessibility menu, dialog, popup

UserName

CAS RN 170899-08-8

IUPAC Name: (2R)-2-{[(tert-butoxy)carbonyl]amino}pent-4-enoate
Synonyms: (2R)-2-[(tert-butoxycarbonyl)amino]pent-4-enoate
Molecular Weight (g/mol): 214.24
Molecular Formula: C10H16NO4
InChi Key: BUPDPLXLAKNJMI-SSDOTTSWSA-M
SMILES: CC(C)(C)OC(=O)N[C@H](CC=C)C([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)