missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-BOC-Allylglycine, 95%, 98% ee

Catalog No. p-69811
Change view
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC362845000 500 mg Glass bottle
AC362840010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC362845000 Supplier Thermo Scientific Chemicals Supplier No. 362845000
Only null left

CAS: 170899-08-8 | C10H17NO4 | 215.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170899-08-8
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.25
MDL Number MFCD01320882
InChI Key BUPDPLXLAKNJMI-SSDOTTSWSA-N
Synonym boc-d-allylglycine, r-2-tert-butoxycarbonyl amino pent-4-enoic acid, boc-alpha-allyl-d-gly, boc-d-allyglycine.dcha, 2r-2-tert-butoxycarbonyl amino pent-4-enoic acid, 4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2r, boc-d-allylglycine.dcha, d-boc-allylglycine, boc-d-allyglycine?dcha, boc-d-allyglycine?cha
PubChem CID 638723
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
SMILES CC(C)(C)OC(=O)NC(CC=C)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Enantiomeric Excess 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 500 mg
Specific Rotation -9.5° to -12.5° (20°C, 589nm) (c=1, methanol)
Formula Weight 215.25
Percent Purity 95%,98% ee
Physical Form Viscous Liquid
Chemical Name or Material (R)-N-BOC-Allylglycine, 95%, (98% e.e.)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
TSCA TSCA
Recommended Storage 4°C

RUO – Research Use Only