missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-N-(3,5-Dinitrobenzoyl)-alpha-phenylglycine 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 345.27 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (R)-(-)-N-(3,5-Dinitrobenzoyl)-alpha-phenylglycine |
Safety and Handling
| TSCA | No |