CAS RN 74927-72-3
IUPAC Name:
(2R)-2-[(3,5-dinitrophenyl)formamido]-2-phenylacetate
Synonyms:
(2R)-2-[(3,5-dinitrophenyl)formamido]-2-phenylacetate
Molecular Weight (g/mol):
344.26
Molecular Formula:
C15H10N3O7
InChi Key:
MIVUDAUOXJDARR-CYBMUJFWSA-M
SMILES:
[O-]C(=O)[C@H](NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O)C1=CC=CC=C1
1
–
1
of
1
results
1
(R)-(-)-N-(3,5-Dinitrobenzoyl)-alpha-phenylglycine 98.0+%, TCI America™
CAS: 74927-72-3 Molecular Formula: C15H10N3O7 Molecular Weight (g/mol): 344.26 MDL Number: MFCD00010134,MFCD00010134,MFCD00064470 InChI Key: MIVUDAUOXJDARR-CYBMUJFWSA-M PubChem CID: 126504 IUPAC Name: (2R)-2-[(3,5-dinitrophenyl)formamido]-2-phenylacetate SMILES: [O-]C(=O)[C@H](NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O)C1=CC=CC=C1