missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-N-(3,5-Dinitrobenzoyl)-alpha-phenylglycine 98.0+%, TCI America™

Catalog No. D18531G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D18531G 1 g
D18535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D18531G Supplier TCI America Supplier No. D18531G
Only null left

Chemical Identifiers

CAS 74927-72-3
Molecular Formula C15H10N3O7
Molecular Weight (g/mol) 344.26
MDL Number MFCD00010134,MFCD00010134,MFCD00064470
InChI Key MIVUDAUOXJDARR-CYBMUJFWSA-M
PubChem CID 126504
IUPAC Name (2R)-2-[(3,5-dinitrophenyl)formamido]-2-phenylacetate
SMILES [O-]C(=O)[C@H](NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 345.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-N-(3,5-Dinitrobenzoyl)-alpha-phenylglycine