missing translation for 'onlineSavingsMsg'
Learn More

(R)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide 98.0+%, TCI America™

Catalog No. b55181g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55181G 1 g
B55185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55181G Supplier TCI America Supplier No. B55181G
Only null left

Chemical Identifiers

CAS 105024-93-9
Molecular Formula C25H24N2O2
Molecular Weight (g/mol) 384.479
MDL Number MFCD18910207
InChI Key IPSABLMEYFYEHS-HSZRJFAPSA-N
PubChem CID 980838
IUPAC Name (2R)-N-(2-benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
SMILES C1CC(N(C1)CC2=CC=CC=C2)C(=O)NC3=CC=CC=C3C(=O)C4=CC=CC=C4

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 1 g
Formula Weight 384.48
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-N-(2-Benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide