accessibility menu, dialog, popup

UserName

NONHO

CAS RN 105024-93-9

IUPAC Name: N-(2-benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
Synonyms: N-(2-benzoylphenyl)-1-benzylpyrrolidine-2-carboxamide
Molecular Weight (g/mol): 384.48
Molecular Formula: C25H24N2O2
InChi Key: IPSABLMEYFYEHS-UHFFFAOYNA-N
SMILES: O=C(NC1=CC=CC=C1C(=O)C1=CC=CC=C1)C1CCCN1CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results