missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-Limonene, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
(R)-(+)-Limonene is a terpenoid compound that is abundantly present in many plants such as rosemary. It is highly associated with plant and insect interactions due to its tendency to attract insects.
$112.64 - $1416.33
Specifications
| CAS | 5989-27-5 |
|---|---|
| Vapor Pressure | <3 mmHg (14.4°C) |
| Vapor Density | 4.7 (vs. air) |
| Density | 0.842 g/mL (at 20°C) |
| Flash Point | 51°C |
Description
Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal CareSpecifications
| 5989-27-5 | |
| 4.7 (vs. air) | |
| 51°C | |
| C10H16 | |
| 176°C to 177°C | |
| Analytical Standard | |
| ≥99% (sum of enantiomers, GC) |
| <3 mmHg (14.4°C) | |
| 0.842 g/mL (at 20°C) | |
| Neat | |
| MFCD00062991 | |
| (+)-p-Mentha-1,8-diene; (+)-Carvene; (R)-4-Isopropenyl-1-methyl-1-cyclohexene | |
| 136.23 | |
| Analytical Standard |