missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-Limonene, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111020239
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020239 1 mL
111018775 5 mL
111018030 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111020239 Supplier Merck Emd Millipore Supplier No. 621181ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(R)-(+)-Limonene is a terpenoid compound that is abundantly present in many plants such as rosemary. It is highly associated with plant and insect interactions due to its tendency to attract insects.

Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 5989-27-5
Vapor Pressure <3 mmHg (14.4°C)
Vapor Density 4.7 (vs. air)
Density 0.842 g/mL (at 20°C)
Flash Point 51°C
Physical Form Neat
Linear Formula C10H16
MDL Number MFCD00062991
Boiling Point 176°C to 177°C
Synonym (+)-p-Mentha-1,8-diene; (+)-Carvene; (R)-4-Isopropenyl-1-methyl-1-cyclohexene
Type Analytical Standard
Quantity 1 mL
Formula Weight 136.23
Percent Purity ≥99% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less