missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DSR 6434

Catalog No. 480950
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
480950 50 mg
480910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 480950 Supplier Tocris Bioscience™ Supplier No. 4809/50

Potent TLR7 agonist

DSR 6434 is a potent TLR7 agonist (EC50 = 7.2 nM). Activates several immune effector cells in vitro. Suppresses metastasis in vivo and enhances efficacy of radiation therapy in a model of colorectal carcinoma.

Specifications

Chemical Name or Material DSR 6434
CAS 1059070-10-8
Synonym 6-Amino-2-(butylamino)-9-[[6-[2-(dimethylamino)ethoxy]-3-pyridinyl]methyl]-7,9-dihydro-8H-purin-8-one
Quantity 50 mg
Solubility Soluble to 50mM in 1eq. HCl and to 100mM in DMSO
Molecular Formula C19H28N8O2
Formula Weight 400.48
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use