missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cerestat

Catalog No. 449250
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
449250 50 mg
449210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 449250 Supplier Tocris Bioscience™ Supplier No. 4492/50

Potent and noncompetitive NMDA receptor antagonist

Cerestat is a potent and noncompetitive NMDA receptor antagonist (IC50 = 36 nM). Exhibits >70-fold selectivity for NMDA receptor over σ receptor. Neuroprotective in a rat model of focal ischemia.

Specifications

Chemical Name or Material Cerestat
CAS 137160-11-3
Synonym N-(3-Ethylphenyl)-N-methyl-N'-1-naphthalenylguanidine hydrochloride
Quantity 50 mg
Solubility Soluble to 50mM in water and to 100mM in DMSO
Molecular Formula C20H21N3·HCl
Formula Weight 339.86
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use