missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(+)-beta-Methylphenethylamine 98.0+%, TCI America™
Supplier: TCI America M17465G
Specifications
| (R)-(+)-beta-Methylphenethylamine | |
| Yellow | |
| C9H13N | |
| 5 g | |
| (R)-(+)-2-Phenyl-1-propylamine | |
| CC(CN)C1=CC=CC=C1 | |
| 135.21 | |
| 135.21 | |
| Liquid |
| 28163-64-6 | |
| 102°C | |
| MFCD00216740 | |
| 2735 | |
| AXORVIZLPOGIRG-QMMMGPOBSA-N | |
| (2R)-2-phenylpropan-1-amine | |
| 1547949 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 28163-64-6 | |
| 135.21 | |
| AXORVIZLPOGIRG-QMMMGPOBSA-N | |
| 1547949 | |
| CC(CN)C1=CC=CC=C1 |
| C9H13N | |
| MFCD00216740 | |
| (R)-(+)-2-Phenyl-1-propylamine | |
| (2R)-2-phenylpropan-1-amine |
Safety and Handling
TSCA : No