missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-beta-Methylphenethylamine 98.0+%, TCI America™

Catalog No. p-7132836
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17465G 5 g
M174625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17465G Supplier TCI America Supplier No. M17465G

Chemical Identifiers

CAS 28163-64-6
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00216740
InChI Key AXORVIZLPOGIRG-QMMMGPOBSA-N
Synonym (R)-(+)-2-Phenyl-1-propylamine
PubChem CID 1547949
IUPAC Name (2R)-2-phenylpropan-1-amine
SMILES CC(CN)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 102°C
Quantity 5 g
UN Number 2735
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-beta-Methylphenethylamine