missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-alpha-Methoxyphenylacetic Acid 98.0+%, TCI America™
$34.66 - $236.80
Chemical Identifiers
| CAS | 3966-32-3 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.176 |
| MDL Number | MFCD00004250 |
| InChI Key | DIWVBIXQCNRCFE-MRVPVSSYSA-N |
| Synonym | r---alpha-methoxyphenylacetic acid, r-2-methoxy-2-phenylacetic acid, 2r-2-methoxy-2-phenylacetic acid, r-methoxyphenylacetic acid, r-methoxy phenyl acetic acid, methoxy phenyl acetic acid #, r---.alpha.-methoxyphenylacetic acid, benzeneacetic acid, .alpha.-methoxy-, r, o-methyl-d-mandelic acid, ksc496q9j |
| PubChem CID | 2724294 |
| IUPAC Name | (2R)-2-methoxy-2-phenylacetic acid |
| SMILES | COC(C1=CC=CC=C1)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M0830100MG
|
TCI America
M0830100MG |
100 mg |
Each for $34.66
|
|
|||||
|
M08301G
|
TCI America
M08301G |
1 g |
Each for $75.08
|
|
|||||
|
M08305G
|
TCI America
M08305G |
5 g |
Each for $236.80
|
|
|||||
Chemical Identifiers
| 3966-32-3 | |
| 166.176 | |
| DIWVBIXQCNRCFE-MRVPVSSYSA-N | |
| 2724294 | |
| COC(C1=CC=CC=C1)C(=O)O |
| C9H10O3 | |
| MFCD00004250 | |
| r---alpha-methoxyphenylacetic acid, r-2-methoxy-2-phenylacetic acid, 2r-2-methoxy-2-phenylacetic acid, r-methoxyphenylacetic acid, r-methoxy phenyl acetic acid, methoxy phenyl acetic acid #, r---.alpha.-methoxyphenylacetic acid, benzeneacetic acid, .alpha.-methoxy-, r, o-methyl-d-mandelic acid, ksc496q9j | |
| (2R)-2-methoxy-2-phenylacetic acid |
Specifications
| 3966-32-3 | |
| White | |
| C9H10O3 | |
| 100 mg | |
| DIWVBIXQCNRCFE-MRVPVSSYSA-N | |
| (2R)-2-methoxy-2-phenylacetic acid | |
| 2724294 | |
| ≥98.0% (GC,T) | |
| (R)-(-)-alpha-Methoxyphenylacetic Acid |
| 68°C | |
| 130°C | |
| MFCD00004250 | |
| r---alpha-methoxyphenylacetic acid, r-2-methoxy-2-phenylacetic acid, 2r-2-methoxy-2-phenylacetic acid, r-methoxyphenylacetic acid, r-methoxy phenyl acetic acid, methoxy phenyl acetic acid #, r---.alpha.-methoxyphenylacetic acid, benzeneacetic acid, .alpha.-methoxy-, r, o-methyl-d-mandelic acid, ksc496q9j | |
| COC(C1=CC=CC=C1)C(=O)O | |
| 166.176 | |
| 166.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No