missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-alpha-Methoxyphenylacetic Acid 98.0+%, TCI America™
Supplier: TCI America M08301G
Specifications
| (R)-(-)-alpha-Methoxyphenylacetic Acid | |
| 68°C | |
| 130°C | |
| MFCD00004250 | |
| r---alpha-methoxyphenylacetic acid, r-2-methoxy-2-phenylacetic acid, 2r-2-methoxy-2-phenylacetic acid, r-methoxyphenylacetic acid, r-methoxy phenyl acetic acid, methoxy phenyl acetic acid #, r---.alpha.-methoxyphenylacetic acid, benzeneacetic acid, .alpha.-methoxy-, r, o-methyl-d-mandelic acid, ksc496q9j | |
| COC(C1=CC=CC=C1)C(=O)O | |
| 166.176 | |
| 166.18 | |
| Crystalline Powder |
| 3966-32-3 | |
| White | |
| C9H10O3 | |
| 1 g | |
| DIWVBIXQCNRCFE-MRVPVSSYSA-N | |
| (2R)-2-methoxy-2-phenylacetic acid | |
| 2724294 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 3966-32-3 | |
| 166.176 | |
| DIWVBIXQCNRCFE-MRVPVSSYSA-N | |
| 2724294 | |
| COC(C1=CC=CC=C1)C(=O)O |
| C9H10O3 | |
| MFCD00004250 | |
| r---alpha-methoxyphenylacetic acid, r-2-methoxy-2-phenylacetic acid, 2r-2-methoxy-2-phenylacetic acid, r-methoxyphenylacetic acid, r-methoxy phenyl acetic acid, methoxy phenyl acetic acid #, r---.alpha.-methoxyphenylacetic acid, benzeneacetic acid, .alpha.-methoxy-, r, o-methyl-d-mandelic acid, ksc496q9j | |
| (2R)-2-methoxy-2-phenylacetic acid |
Safety and Handling
TSCA : No