missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-9-(2-Hydroxypropyl)adenine 98.0+%, TCI America™
Supplier: TCI America H12915G
Specifications
| (R)-9-(2-Hydroxypropyl)adenine | |
| 200°C | |
| C8H11N5O | |
| 5 g | |
| MJZYTEBKXLVLMY-UHFFFAOYNA-N | |
| 1-(6-amino-9H-purin-9-yl)propan-2-ol | |
| 445211 | |
| ≥98.0% (HPLC,T) |
| 14047-28-0 | |
| White-Yellow | |
| MFCD07369451 | |
| (R)-6-Amino-9-(2-hydroxypropyl)purine | |
| CC(O)CN1C=NC2=C(N)N=CN=C12 | |
| 193.21 | |
| 193.21 | |
| Crystalline Powder |
Chemical Identifiers
| 14047-28-0 | |
| 193.21 | |
| MJZYTEBKXLVLMY-UHFFFAOYNA-N | |
| 445211 | |
| CC(O)CN1C=NC2=C(N)N=CN=C12 |
| C8H11N5O | |
| MFCD07369451 | |
| (R)-6-Amino-9-(2-hydroxypropyl)purine | |
| 1-(6-amino-9H-purin-9-yl)propan-2-ol |
Safety and Handling
TSCA : No