missing translation for 'onlineSavingsMsg'
Learn More

(R)-9-(2-Hydroxypropyl)adenine 98.0+%, TCI America™

Catalog No. p-7130963
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12911G 1 g
H12915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12911G Supplier TCI America Supplier No. H12911G

Chemical Identifiers

CAS 14047-28-0
Molecular Formula C8H11N5O
Molecular Weight (g/mol) 193.21
MDL Number MFCD07369451
InChI Key MJZYTEBKXLVLMY-UHFFFAOYNA-N
Synonym (R)-6-Amino-9-(2-hydroxypropyl)purine
PubChem CID 445211
IUPAC Name 1-(6-amino-9H-purin-9-yl)propan-2-ol
SMILES CC(O)CN1C=NC2=C(N)N=CN=C12

Specifications

Melting Point 200°C
Color White-Yellow
Quantity 1 g
Formula Weight 193.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-9-(2-Hydroxypropyl)adenine