accessibility menu, dialog, popup

UserName

H3CNNNNNH2HO

CAS RN 14047-28-0

IUPAC Name: 1-(6-amino-9H-purin-9-yl)propan-2-ol
Synonyms: 1-(6-aminopurin-9-yl)propan-2-ol
Molecular Weight (g/mol): 193.21
Molecular Formula: C8H11N5O
InChi Key: MJZYTEBKXLVLMY-UHFFFAOYNA-N
SMILES: CC(O)CN1C=NC2=C(N)N=CN=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results