missing translation for 'onlineSavingsMsg'
Learn More

(R)-5,5-Diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine 97.0+%, TCI America™

Catalog No. p-7127123
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D21301G 1 g
D21305G 5 g
2 options

Catalog No. D21301G

Supplier: TCI America D21301G

Only null left

Chemical Identifiers

CAS 112022-83-0
Molecular Formula C18H20BNO
Molecular Weight (g/mol) 277.17
MDL Number MFCD00078440
InChI Key VMKAFJQFKBASMU-QGZVFWFLSA-N
Synonym r-2-methyl-cbs-oxazaborolidine, r-methyl oxazaborolidine, r-+-2-methyl-cbs-oxazaborolidine, r-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, r-1-methyl-3,3-diphenylhexahydropyrrolo 1,2-c 1,3,2 oxazaborole, r-me-cbs catalyst, r-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, r-me cbs, r-tetrahydro-1-methyl-3,3-diphenyl-1h,3h-pyrrolo 1,2-c 1,3,2 oxazaborole, corey's catalyst
PubChem CID 9838490
IUPAC Name (3aR)-1-methyl-3,3-diphenyl-hexahydropyrrolo[1,2-c][1,3,2]oxazaborole
SMILES [H][C@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 277.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-5,5-Diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine