missing translation for 'onlineSavingsMsg'
Learn More

(R)-4-(1-Aminoethyl)phenol, 97%

Catalog No. AAH6300103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6300103 1 g
AAH6300106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6300103 Supplier Thermo Scientific Chemicals Supplier No. H6300103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134855-89-3 | C8H12BrNO | 218.094 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 134855-89-3
Molecular Formula C8H12BrNO
Molecular Weight (g/mol) 218.094
MDL Number MFCD03844647
InChI Key PZBBMKOZPQAHRA-RGMNGODLSA-N
Synonym s-4-1-amino-ethyl-phenol hydrobromid, s---1-4-hydroxyphenyl ethylamine hydrobromide
PubChem CID 49758827
IUPAC Name 4-[(1S)-1-aminoethyl]phenol;hydrobromide
SMILES CC(C1=CC=C(C=C1)O)N.Br

Specifications

Color White
Quantity 1 g
UN Number UN3259
Formula Weight 137.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (R)-4-(1-Aminoethyl)phenol
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only