CAS RN 134855-89-3
IUPAC Name:
4-(1-aminoethyl)phenol hydrobromide
Synonyms:
1-(p-hydroxyphenyl)ethylamine hydrobromide
Molecular Weight (g/mol):
218.09
Molecular Formula:
C8H12BrNO
InChi Key:
PZBBMKOZPQAHRA-UHFFFAOYNA-N
SMILES:
Br.CC(N)C1=CC=C(O)C=C1
1
–
1
of
1
results
1
Promotions Available
(R)-4-(1-Aminoethyl)phenol, 97%
(R)-4-(1-Aminoethyl)phenol, 97%
CAS: 134855-89-3 Molecular Formula: C8H12BrNO Molecular Weight (g/mol): 218.094 MDL Number: MFCD03844647 InChI Key: PZBBMKOZPQAHRA-RGMNGODLSA-N Synonym: s-4-1-amino-ethyl-phenol hydrobromid,s---1-4-hydroxyphenyl ethylamine hydrobromide PubChem CID: 49758827 IUPAC Name: 4-[(1S)-1-aminoethyl]phenol;hydrobromide SMILES: CC(C1=CC=C(C=C1)O)N.Br