missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-3-(tert-Butoxycarbonylamino)piperidine 98.0+%, TCI America™
$70.25 - $231.64
Chemical Identifiers
| CAS | 309956-78-3 |
|---|---|
| Molecular Formula | C10H20N2O2 |
| Molecular Weight (g/mol) | 200.282 |
| MDL Number | MFCD03093382 |
| InChI Key | WUOQXNWMYLFAHT-MRVPVSSYSA-N |
| Synonym | r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester |
| PubChem CID | 1514172 |
| IUPAC Name | tert-butyl N-[(3R)-piperidin-3-yl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CCCNC1 |
Chemical Identifiers
| 309956-78-3 | |
| 200.282 | |
| WUOQXNWMYLFAHT-MRVPVSSYSA-N | |
| 1514172 | |
| CC(C)(C)OC(=O)NC1CCCNC1 |
| C10H20N2O2 | |
| MFCD03093382 | |
| r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester | |
| tert-butyl N-[(3R)-piperidin-3-yl]carbamate |
Specifications
| 123°C | |
| White-Yellow | |
| MFCD03093382 | |
| WUOQXNWMYLFAHT-MRVPVSSYSA-N | |
| tert-butyl N-[(3R)-piperidin-3-yl]carbamate | |
| 1514172 | |
| ≥98.0% (GC,T) | |
| (R)-3-(tert-Butoxycarbonylamino)piperidine |
| 309956-78-3 | |
| C10H20N2O2 | |
| r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester | |
| CC(C)(C)OC(=O)NC1CCCNC1 | |
| 200.282 | |
| 200.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No