missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-(tert-Butoxycarbonylamino)piperidine 98.0+%, TCI America™

Catalog No. B35911G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35911G 1 g
B35915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35911G Supplier TCI America Supplier No. B35911G

Chemical Identifiers

CAS 309956-78-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03093382
InChI Key WUOQXNWMYLFAHT-MRVPVSSYSA-N
Synonym r-3-boc-aminopiperidine, r-3-boc-amino piperidine, r-tert-butyl piperidin-3-ylcarbamate, tert-butyl n-3r-piperidin-3-yl carbamate, tert-butyl r-piperidin-3-ylcarbamate, r-3-tert-butoxycarbonylamino piperidine, r-3-boc-amino-piperidine, r-+-3-tert-butoxycarbonylaminopiperidine, r-piperidin-3-yl-carbamic acid tert-butyl ester
PubChem CID 1514172
IUPAC Name tert-butyl N-[(3R)-piperidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCNC1

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 1 g
Formula Weight 200.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-3-(tert-Butoxycarbonylamino)piperidine