missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine, 97%, Thermo Scientific Chemicals

$1632.90 - $1632.90

Chemical Identifiers

CAS 1212136-00-9
Molecular Formula C19H21NO7S
Molecular Weight (g/mol) 407.437
MDL Number MFCD09864994
InChI Key RTHSATBSXDKZBN-QGZVFWFLSA-N
Synonym methyl r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2,3 oxathiazolidine-4-carboxylate, methyl 4r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2??,3-oxathiazolidine-4-carboxylate
PubChem CID 46172874
IUPAC Name methyl (4R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxooxathiazolidine-4-carboxylate
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)N3C(COS3(=O)=O)C(=O)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6388906
View Documents
Thermo Scientific Chemicals
H6388906
5 g
Each for $1,632.90
Only null left
 
Description

Description

Thiazoles are used in peptide research. Mainly used in medicine field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazoles are used in peptide research. Mainly used in medicine field.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. 2-7°C Refrigerate.
Specifications

Chemical Identifiers

1212136-00-9
407.437
RTHSATBSXDKZBN-QGZVFWFLSA-N
46172874
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)N3C(COS3(=O)=O)C(=O)OC
C19H21NO7S
MFCD09864994
methyl r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2,3 oxathiazolidine-4-carboxylate, methyl 4r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2??,3-oxathiazolidine-4-carboxylate
methyl (4R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxooxathiazolidine-4-carboxylate

Specifications

1212136-00-9
White to Brown
MFCD09864994
methyl r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2,3 oxathiazolidine-4-carboxylate, methyl 4r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2??,3-oxathiazolidine-4-carboxylate
COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)N3C(COS3(=O)=O)C(=O)OC
407.437
237.27
Solid
100°C to 107°C
C19H21NO7S
5 g
RTHSATBSXDKZBN-QGZVFWFLSA-N
methyl (4R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxooxathiazolidine-4-carboxylate
46172874
97%
(R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only