accessibility menu, dialog, popup

UserName

CAS RN 1212136-00-9

IUPAC Name: methyl 3-[bis(4-methoxyphenyl)methyl]-2,2-dioxo-1,2λ⁶,3-oxathiazolidine-4-carboxylate
Synonyms: methyl 3-[bis(4-methoxyphenyl)methyl]-2,2-dioxo-1,2λ⁶,3-oxathiazolidine-4-carboxylate
Molecular Weight (g/mol): 407.44
Molecular Formula: C19H21NO7S
InChi Key: RTHSATBSXDKZBN-UHFFFAOYNA-N
SMILES: COC(=O)C1COS(=O)(=O)N1C(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results