missing translation for 'onlineSavingsMsg'
Learn More

(R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine, 97%, Thermo Scientific Chemicals

Catalog No. AAH6388906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6388906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6388906 Supplier Thermo Scientific Chemicals Supplier No. H6388906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1212136-00-9 | Molecular Formula: C19H21NO7S | Molecular Weight: 407.437 g/mol

Thiazoles are used in peptide research. Mainly used in medicine field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiazoles are used in peptide research. Mainly used in medicine field.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. 2-7°C Refrigerate.

Chemical Identifiers

CAS 1212136-00-9
Molecular Formula C19H21NO7S
Molecular Weight (g/mol) 407.437
MDL Number MFCD09864994
InChI Key RTHSATBSXDKZBN-QGZVFWFLSA-N
Synonym methyl r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2,3 oxathiazolidine-4-carboxylate, methyl 4r-3-bis 4-methoxyphenyl methyl-2,2-dioxo-1,2??,3-oxathiazolidine-4-carboxylate
PubChem CID 46172874
IUPAC Name methyl (4R)-3-[bis(4-methoxyphenyl)methyl]-2,2-dioxooxathiazolidine-4-carboxylate
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)N3C(COS3(=O)=O)C(=O)OC

Specifications

Melting Point 100°C to 107°C
Color White to Brown
Quantity 5 g
Formula Weight 237.27
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (R)-3-Boc-4-methyl-2,2-dioxo-1,2,3-oxathiazolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only