missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-2-Phenylglycinol 98.0+%, TCI America™
Supplier: TCI America P120125G
Specifications
| (R)-(-)-2-Phenylglycinol | |
| 79°C | |
| C8H12NO | |
| 25 g | |
| IJXJGQCXFSSHNL-MRVPVSSYSA-O | |
| (1S)-2-hydroxy-1-phenylethan-1-aminium | |
| 2724025 | |
| ≥98.0% (GC,T) |
| 56613-80-0 | |
| White-Yellow | |
| MFCD00008062 | |
| r---2-phenylglycinol, d-phenylglycinol, r-2-phenylglycinol, r-2-amino-2-phenylethanol, 2r-2-amino-2-phenylethan-1-ol, d-plenylglycinol, r---2-amino-2-phenylethanol, 2r-2-amino-2-phenylethanol, d-2-phenylglycinol, r-phenylglycinol | |
| [NH3+][C@H](CO)C1=CC=CC=C1 | |
| 138.19 | |
| 137.18 | |
| Crystalline Powder |
Chemical Identifiers
| 56613-80-0 | |
| 138.19 | |
| IJXJGQCXFSSHNL-MRVPVSSYSA-O | |
| 2724025 | |
| [NH3+][C@H](CO)C1=CC=CC=C1 |
| C8H12NO | |
| MFCD00008062 | |
| r---2-phenylglycinol, d-phenylglycinol, r-2-phenylglycinol, r-2-amino-2-phenylethanol, 2r-2-amino-2-phenylethan-1-ol, d-plenylglycinol, r---2-amino-2-phenylethanol, 2r-2-amino-2-phenylethanol, d-2-phenylglycinol, r-phenylglycinol | |
| (1S)-2-hydroxy-1-phenylethan-1-aminium |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator