missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2-Phenylglycinol 98.0+%, TCI America™

Catalog No. p-7135005
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12015G 5 g
P120125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12015G Supplier TCI America Supplier No. P12015G
Only null left

Chemical Identifiers

CAS 56613-80-0
Molecular Formula C8H12NO
Molecular Weight (g/mol) 138.19
MDL Number MFCD00008062
InChI Key IJXJGQCXFSSHNL-MRVPVSSYSA-O
Synonym r---2-phenylglycinol, d-phenylglycinol, r-2-phenylglycinol, r-2-amino-2-phenylethanol, 2r-2-amino-2-phenylethan-1-ol, d-plenylglycinol, r---2-amino-2-phenylethanol, 2r-2-amino-2-phenylethanol, d-2-phenylglycinol, r-phenylglycinol
PubChem CID 2724025
IUPAC Name (1S)-2-hydroxy-1-phenylethan-1-aminium
SMILES [NH3+][C@H](CO)C1=CC=CC=C1

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 5 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-2-Phenylglycinol
TSCA No
Recommended Storage Refrigerator